missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MeCP2, p Ser80 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-47558
This item is not returnable.
View return policy
Description
MeCP2 Polyclonal specifically detects MeCP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MeCP2 (p Ser80) | |
| Polyclonal | |
| Western Blot 1:100 - 1:1000, Immunohistochemistry 1:100, Immunohistochemistry-Paraffin 1:100 | |
| P51608 | |
| MECP2 | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser80 conjugated to KLH. Accession # P51608 | |
| Unpurified | |
| RUO | |
| Primary | |
| Specific for the ∼75 kDa MECP2 protein phosphorylated at Ser80 in Western blots of human, rat and mouse brain extracts. The antibody has also been used successfully for immunohistochemistry on mouse brain sections. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
| AUTSX3, DKFZp686A24160, MeCp-2 protein, mental retardation, X-linked 79, methyl CpG binding protein 2 (Rett syndrome), X-linked 16 | |
| Rabbit | |
| 75 kDa | |
| 0.1 mL | |
| Cancer, Chromatin Research, Neuroscience, Phospho Specific | |
| 4204 | |
| Human, Mouse, Rat | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction