Learn More
Invitrogen™ MECP2 Polyclonal Antibody
GREENER_CHOICE
Chicken Polyclonal Antibody
Brand:  Invitrogen™ PA5143577
 View more versions of this product
Product Code. 17846417
  • € 491.00 / 100µL
Description
MECP2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IHC (PFA)
MECP2 belongs to a family of nuclear proteins (including MBD1, MBD2, MBD3 and MBD4) that have a methyl-CpG binding domain. MECP2 is capable of binding specifically to methylated DNA. DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. MECP2 can also repress transcription from methylated gene promoters. In contrast to other MBD family members, MECP2 is X-linked and subject to X inactivation. MECP2 is dispensible in stem cells, but is essential for embryonic development. MECP2 gene mutations are the cause of most cases of Rett syndrome, a progressive neurologic developmental disorder and one of the most common causes of mental retardation in females.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MECP2
Polyclonal
Unconjugated
Mecp2
1500041B07Rik; AUTSX3; D630021H01Rik; DKFZp686A24160; Mbd5; MECP2; MeCp-2 protein; methyl CpG binding protein 2; methyl-CpG binding protein 2; methyl-CpG-binding protein 2; MRX16; MRX79; MRXS13; MRXSL; PPMX; RS; RTS; RTT; testis tissue sperm-binding protein Li 41a; WBP10
Chicken
100 μL
Primary
Human, Mouse, Rat, Monkey
Antibody
IgY
Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry
Conc. Not Determined
PBS with 5mM sodium azide
P51608, Q00566, Q9Z2D6
Mecp2
Full length recombinant human MeCP2 expressed in and purified from E. coli.
RUO
17257, 29386, 4204
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.