missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MECR Polyclonal Antibody
GREENER_CHOICE

Product Code. 15782481 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15782481 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15782481 Supplier Invitrogen™ Supplier No. PA554553

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: SDIPLQSAAT LGVNPCTAYR MLMDFEQLQP GDSVIQNASN SGVGQAVIQI AAALGLRTIN VVRDRPDIQK LSDRLKSLGA EHVITEEELR RPEMKNFFKD MPQPRLALNC V Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.

The mitochondrial trans-2-enoyl-CoA reductase (MECR), was initially identified as nuclear receptor-binding factor 1 (NRBF1), which can interact with a multitude of nuclear hormone receptors in the presence of the respective ligands. MECR has been shown to be part of the mitochondrial fatty acid synthesis (FAS II) system and to catalyze the NAPDH-dependent reduction of 2-enoyl thioesters, generating saturated acyl-groups. Overexpression of this gene in transgenic mice can lead to cardiac abnormalities, suggesting that inappropriate expression of genes of FAS II can result in the development of hereditary cardiomyopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MECR
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mecr
Gene Accession No. Q9BV79
Gene Alias 2-enoyl thioester reductase; AI195831; CGI-63; Enoyl-[acyl-carrier-protein] reductase, mitochondrial; FASN2B; homolog of yeast 2-enoyl thioester reductase; HsNrbf-1; Mecr; mitochondrial 2-enoyl thioester reductase; mitochondrial trans-2-enoyl-CoA reductase; NBRF1; Nrbf1; NRBF-1; nuclear receptor binding factor 1; nuclear receptor-binding factor 1; trans-2-enoyl-CoA reductase, mitochondrial
Gene Symbols Mecr
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MECR. Recombinant protein control fragment (Product #RP-93135).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51102
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.