missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED14 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-83187-0.1ML
This item is not returnable.
View return policy
Description
MED14 Polyclonal specifically detects MED14 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation.
Specifications
| MED14 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ChIP Assay | |
| Unconjugated | |
| Activator-recruited cofactor 150 kDa component, ARC150, Cofactor required for Sp1 transcriptional activation subunit 2, cofactor required for Sp1 transcriptional activation, subunit 2 (150kD), CRSP150, CRSP2cofactor required for Sp1 transcriptional activation, subunit 2, 150kDa, CSRP, CXorf4MGC104513, EXLM1CRSP complex subunit 2, human homolog of yeast RGR1, mediator complex subunit 14DRIP150, mediator of RNA polymerase II transcription subunit 14, RGR1RGR1 homolog, Thyroid hormone receptor-associated protein complex 170 kDa component, thyroid hormone receptor-associated protein complex component TRAP170, transcriptional co-activator CRSP150, Transcriptional coactivator CRSP150, Trap170, TRAP170hRGR1, vitamin D receptor-interacting protein complex component DRIP150, Vitamin D3 receptor-interacting protein complex 150 kDa component | |
| The immunogen is a synthetic peptide directed towards the middle region of human MED14 | |
| 0.1 mL | |
| Signal Transduction | |
| 9282 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction