missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED15 Antibody (OTI1C11), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72618R
This item is not returnable.
View return policy
Description
MED15 Monoclonal specifically detects MED15 in Human samples. It is validated for Immunohistochemistry.
Specifications
| MED15 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MED15 | |
| Recombinant protein fragment corresponding to amino acids 393-679 of human MED15 (NP_056973) produced in E.coli. | |
| 0.1 mL | |
| 51586.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI1C11 | |
| Immunohistochemistry | |
| ARC105Arc105, CAG7A, CTG repeat protein 7a, CTG7A, FLJ42282, FLJ42935, mediator complex subunit 15PC2 glutamine/Q-rich-associated protein, mediator of RNA polymerase II transcription subunit 15, PC2 (positive cofactor 2, multiprotein complex) glutamine/Q-rich-associatedprotein, PC2-glutamine-rich-associated protein, PCQAPDKFZp686A2214, Positive cofactor 2 glutamine/Q-rich-associated protein, positive cofactor 2, glutamine/Q-rich-associated protein, TIG1DKFZp762B1216, TIG-1TPA-inducible gene 1 protein, TNRC7, TPA inducible gene-1, TPA inducible protein, trinucleotide repeat containing 7,105-kD, Trinucleotide repeat-containing gene 7 protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction