missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED15 Monoclonal Antibody (OTI1C11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807950
This item is not returnable.
View return policy
Description
Component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. Mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors. Required for cholesterol-dependent gene regulation. Positively regulates the Nodal signaling pathway.Specifications
| MED15 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96RN5 | |
| MED15 | |
| Human recombit protein fragment corresponding to amino acids 393-679 of human MED15 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin) | |
| OTI1C11 | |
| Unconjugated | |
| MED15 | |
| ARC105, CAG7A, CTG7A, PCQAP, TIG-1, TIG1, TNRC7 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 51586 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction