missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MED18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551305
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total MED18 protein.
Zinc, an essential element required for cell proliferation and differentiation, plays a role in a diverse array of cellular functions, including acting as a cofactor for numerous enzymes and transcription factors and as a neuroregulator. The zinc transporter (ZnT) family regulates the supply of zinc within cells, and its members are characterized by containing six membrane-spanning domains, a large histidine-rich intracellular loop, and a C-terminal tail. ZnT-1, a ubiquitous protein of approximately 60 kDa, localizes to the plasma membrane to aid in the export of zinc out of cells.
Specifications
| MED18 | |
| Polyclonal | |
| Unconjugated | |
| MED18 | |
| 2810046C01Rik; FLJ20045; Med18; mediator 18; Mediator complex subunit 18; mediator of RNA polymerase II transcription subunit 18; mediator of RNA polymerase II transcription, subunit 18 homolog; p28b; TRAP/mediator complex subunit p28b | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54797 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.7 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9BUE0 | |
| MED18 | |
| Full length fusion protein of human MED18. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction