missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED19 Monoclonal Antibody (OTI3F2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810430S
This item is not returnable.
View return policy
Description
MED19 Monoclonal antibody specifically detects MED19 in Human, Rat samples. It is validated for Western BlotSpecifications
| MED19 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| A0JLT2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 219541, 311165 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3F2 | |
| Unconjugated | |
| MED19 | |
| 2410018M14Rik; 3110040A13Rik; DT2P1G7; LCMR1; Lung cancer metastasis-related protein 1; Med19; MED19AS; mediator complex subunit 19; Mediator of RNA polymerase II transcription subunit 19; mediator of RNA polymerase II transcription, subunit 19 homolog; putative mediator subunit MED19AS protein; RGD1311926 | |
| Full length human recombinant protein of human MED19 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction