missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED19 Mouse anti-Human, Rat, Clone: OTI2E6, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810429
This item is not returnable.
View return policy
Description
MED19 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II.Specifications
| MED19 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| A0JLT2 | |
| MED19 | |
| Full length human recombit protein of human MED19 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2b |
| Western Blot | |
| OTI2E6 | |
| Unconjugated | |
| MED19 | |
| DT2P1G7, LCMR1, MED19AS | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 219541, 311165 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion