missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MED21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568706
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 93%.
This gene encodes a member of the mediator complex subunit 21 family. The encoded protein interacts with the human RNA polymerase II holoenzyme and is involved in transcriptional regulation of RNA polymerase II transcribed genes. A pseudogene of this gene is located on chromosome 8. Alternative splicing results in multiple transcript variants.
Specifications
| MED21 | |
| Polyclonal | |
| Unconjugated | |
| MED21 | |
| 0610007L03Rik; AI449604; D19234; D6Ertd782e; fk48d02; hSrb7; hypothetical protein LOC539496; MED21; Mediator complex subunit 21; mediator of RNA polymerase II transcription subunit 21; RNA polymerase II holoenzyme component SRB7; RNAPII complex component SRB7; si:ch211-203h15.4; SRB7; SRB7 (supressor of RNA polymerase B) homolog; SRB7 suppressor of RNA polymerase B homolog; surb7; wu:fk48d02; zgc:56532 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 108098 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ChIP Assay, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9CQ39 | |
| MED21 | |
| synthetic peptide directed towards the middle region of human MED21 (aa 19-68). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction