missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED23 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB200-337H
This item is not returnable.
View return policy
Description
MED23 Polyclonal antibody specifically detects MED23 in Human, Mouse samples. It is validated for Western Blot
Specifications
| MED23 | |
| Polyclonal | |
| Western Blot | |
| 130 kDa transcriptional co-activator, 133 kDa transcriptional co-activator, CRSP130, DRIP130DKFZp434H0117, mediator complex subunit 23CRSP133, Protein sur-2 homolog, subunit 3, 130kDa, Transcriptional coactivator CRSP130, Vitamin D3 receptor-interacting protein complex 130 kDa component | |
| A synthetic peptide that maps to a region between residues 1 and 50 of human Cofactor Required for Sp1 transcriptional activation, subunit 3 using the numbering given in Swiss-Prot entry Q9ULK4 (GeneID 9439). | |
| 0.1 mL | |
| Cell Biology, Signal Transduction, Transcription Factors and Regulators | |
| 9439 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction