missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED30 Antibody (OTI3B3), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72619AF532
This item is not returnable.
View return policy
Description
MED30 Monoclonal specifically detects MED30 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| MED30 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| MED30 | |
| Full length recombinant protein of human MED30 (NP_542382) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI3B3 | |
| Western Blot | |
| mediator complex subunit 30TRAP/Mediator complex component TRAP25, mediator of RNA polymerase II transcription subunit 30, putative mediator of RNA polymerase II transcription subunit 30, THRAP6MGC9890, thyroid hormone receptor associated protein 6, Thyroid hormone receptor-associated protein 6, Thyroid hormone receptor-associated protein complex 25 kDa component, Trap25, TRAP25MED30S | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 90390.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction