missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED30 Antibody (OTI3B3), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72619DL594
This item is not returnable.
View return policy
Description
MED30 Monoclonal antibody specifically detects MED30 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MED30 | |
| Monoclonal | |
| DyLight 594 | |
| mediator complex subunit 30TRAP/Mediator complex component TRAP25, mediator of RNA polymerase II transcription subunit 30, putative mediator of RNA polymerase II transcription subunit 30, THRAP6MGC9890, thyroid hormone receptor associated protein 6, Thyroid hormone receptor-associated protein 6, Thyroid hormone receptor-associated protein complex 25 kDa component, Trap25, TRAP25MED30S | |
| Full length recombinant protein of human MED30 (NP_542382) produced in E.coli. | |
| 0.1 mL | |
| Signal Transduction | |
| 90390 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot | |
| OTI3B3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction