Learn More
MED30 Mouse anti-Human, Clone: OTI3B3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF808041
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. Mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.Specifications
| MED30 | |
| Monoclonal | |
| Unconjugated | |
| MED30 | |
| MED30S, THRAP6, TRAP25 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 90390 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI3B3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q96HR3 | |
| MED30 | |
| Full length human recombit protein of human MED30 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.