missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MED30 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565417
This item is not returnable.
View return policy
Description
Immunogen sequence: MSTPPLAASGM APGPFAGPQA QQAAREVNTA SLCRIGQETV QDIVYRTMEI FQLLRNMQLP NGVTY Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.
Med30 is a subunit of the RNA polymerase II (Pol II) transcriptional mediator complex. The mediator complex is a coactivator involved in the regulated transcription of Pol II-dependent genes. The mediator complex functions as a bridge to convey information from gene-specific regulatory proteins to the basal Pol II transcription machinery, and is recruited to promoter regions by directly interacting with regulatory proteins. The mediator complex also serves as a scaffold for the assembly of a functional pre-initiation complex with Pol II and other general transcription factors. Found in the nucleus, Med30 is expressed in brain, heart, kidney, liver, lung, pancreas, placenta and skeletal muscle.
Specifications
| MED30 | |
| Polyclonal | |
| Unconjugated | |
| MED30 | |
| 1810038N03Rik; 2510044J04Rik; Med30; MED30S; Mediator complex subunit 30; mediator of RNA polymerase II transcription subunit 30; putative mediator of RNA polymerase II transcription subunit 30; THRAP6; thyroid hormone receptor associated protein 6; thyroid hormone receptor-associated protein 6; Thyroid hormone receptor-associated protein complex 25 kDa component; TRAP/Mediator complex component TRAP25; TRAP25 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90390 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96HR3 | |
| MED30 | |
| Recombinant Human MED30. Recombinant protein control fragment (Product #RP-106097). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction