missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED6 Antibody (OTI3C9), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72620F
This item is not returnable.
View return policy
Description
MED6 Monoclonal antibody specifically detects MED6 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MED6 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 10001 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot | |
| OTI3C9 | |
| Western Blot | |
| Activator-recruited cofactor 33 kDa component, ARC33, mediator complex subunit 6hMed6, mediator of RNA polymerase II transcription subunit 6, mediator of RNA polymerase II transcription, subunit 6 homolog, mediator of RNA polymerase II transcription, subunit 6 homolog (S. cerevisiae), NY-REN-28, Renal carcinoma antigen NY-REN-28 | |
| Full length human recombinant protein of human MED6(NP_005457) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction