missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MED6 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA565079

Product Code. 15917854

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: NSGSVLDYFSE RSNPFYDRTC NNEVVKMQRL TLEHLNQMVG IEYILLHAQE PILFIIRKQQ RQSPAQVIPL ADYYIIAGVI YQAPDLGSVI NSRVLTAVHG IQSAFDEAMS YCRYHPSKGY WWHFKDHEEQ DKVRPKAKRK EEPSSI Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.

MED6 is a component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. And mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MED6
Polyclonal
Unconjugated
Med6
1500012F11Rik; Activator-recruited cofactor 33 kDa component; ARC33; AU019660; AV213303; CTD-2540L5.5; hMed6; hypothetical protein LOC505293; MED6; Mediator complex subunit 6; mediator of RNA polymerase II transcriptio subunit 6-like protein; mediator of RNA polymerase II transcription subunit 6; mediator of RNA polymerase II transcription, subunit 6 homolog; NY-REN-28; Renal carcinoma antigen NY-REN-28
Rabbit
Antigen affinity chromatography
RUO
10001
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O75586
Med6
Recombinant Human MED6. Recombinant protein control fragment (Product #RP-104618).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.