missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MED6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15524442
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15524442 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15524442 Supplier Invitrogen™ Supplier No. PA540846

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: LLLDLRQKFP PKFVQLKPGE KPVPVDQTKK EAEPIPETVK PEEKETTKNV Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 86%; Pig: 93%; Rabbit: 93%; Rat: 93%.

MED6 is a component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. And mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MED6
Applications ChIP Assay, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Med6
Gene Accession No. O75586
Gene Alias 1500012F11Rik; Activator-recruited cofactor 33 kDa component; ARC33; AU019660; AV213303; CTD-2540L5.5; hMed6; hypothetical protein LOC505293; MED6; Mediator complex subunit 6; mediator of RNA polymerase II transcriptio subunit 6-like protein; mediator of RNA polymerase II transcription subunit 6; mediator of RNA polymerase II transcription, subunit 6 homolog; NY-REN-28; Renal carcinoma antigen NY-REN-28
Gene Symbols Med6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human MED6.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10001
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.