missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MED8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555850
This item is not returnable.
View return policy
Description
Immunogen sequence: LLEKISKEER ESESGGLRPN KQTFNPTDTN ALVAAVAFGK GLSNWRPSGS SGPGQAGQPG AGTILAGTSG LQ Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 31%.
This gene encodes a protein that is one of more than 20 subunits of the mediator complex, first identified in S. cerevisiae, that is required for activation of transcription. The product of this gene also interacts with elongins B and C, and CUL2 and RBX1, to reconstitute a ubiquitin ligase. Two alternative transcripts encoding different isoforms have been described.
Specifications
| MED8 | |
| Polyclonal | |
| Unconjugated | |
| Med8 | |
| 2210021A15Rik; AB041805; Activator-recruited cofactor 32 kDa component; ARC32; Med8; Mediator complex subunit 8; mediator of RNA polymerase II transcription subunit 8; mediator of RNA polymerase II transcription subunit MED8; mediator of RNA polymerase II transcription, subunit 8 homolog; MGC17544; MGC19641; MNCb-2386; zgc:76877; zgc:76877 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 112950 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96G25 | |
| Med8 | |
| Recombinant protein corresponding to Human MED8. Recombinant protein control fragment (Product #RP-94337). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction