Learn More
MED9 Monoclonal Antibody (OTI6E3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811890
This item is not returnable.
View return policy
Description
Component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes. Mediator functions as a bridge to convey information from gene-specific regulatory proteins to the basal RNA polymerase II transcription machinery. Mediator is recruited to promoters by direct interactions with regulatory proteins and serves as a scaffold for the assembly of a functional preinitiation complex with RNA polymerase II and the general transcription factors.Specifications
| MED9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NWA0 | |
| MED9 | |
| Full length human recombit protein of human MED9 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI6E3 | |
| Unconjugated | |
| MED9 | |
| MED25 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55090 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |