Learn More
Invitrogen™ MEDAG Polyclonal Antibody
GREENER_CHOICE
Product Code. 15836053
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15836053 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15836053 Supplier Invitrogen™ Supplier No. PA567634
Rabbit Polyclonal Antibody
Predicted to react with Mouse samples.
MEDAG is a protein coding gene involved in processes that promote adipocyte differentiation, lipid accumulation, and glucose uptake in mature adipocytes.
TRUSTED_SUSTAINABILITY
Specifications
Antigen MEDAG
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene MEDAG
Gene Accession No. Q14BA6, Q5VYS4
Gene Alias 6330406I15Rik; Activated in W/Wv mouse stomach 3; activated in W/Wv mouse stomach 3 homolog; Awms3; C12H13orf33; C13orf33; C25H13orf33; hAWMS3; mAWMS3; MEDA4; MEDA-4; Medag; mesenteric estrogen dependent adipogenesis; mesenteric estrogen-dependent adipogenesis; mesenteric estrogen-dependent adipogenesis protein; Mesenteric estrogen-dependent adipose 4; RGD1307396; uncharacterized protein C13orf33 homolog
Gene Symbols MEDAG
Host Species Rabbit
Immunogen Synthesized peptide derived from human C13orf33.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 70717, 84935
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG