Learn More
B Braun™ Medibox 4.7L Waste Bin
Product Code. 15971622
Click to view available options
Capacity (Metric):
3.9 L
Unit Size:
Each
Product Code. 15971622
Brand: B Braun™ 9193618
Edge
A disposal container with a large opening to ease the insertion of various medical sharps.
  • Complying with EN ISO 23907
  • No-touch, twist-off and insertion inlets for all kinds of used luer and luer-lock needles as well as pen needles
  • Easy to use temporary closure
  • Irreversible final lock mechanism
  • Overfill warning by maximum fill line
Specifications
Capacity (Metric) 3.9 L
Closure Type Temporary closure with irreversible locking mechanism
Closure Color White
Color Yellow
Dimensions (L x W x H) 19 x 19 x 20.5 cm
Quantity 25/Pk.
For Use With (Application) Disposal of medical sharps
Product Type Sharps Disposal Box