Learn More
Lenz Laborglasinstrumente™ Medium Shape Weighing Bottles
Click to view available options
Diameter (Metric):
30 mm
38 mm
48 mm
75 mm
Unit Size:
Each
Pack of 6
Specifications
Specifications
| Includes | Interchangeable NS stopper, inscription label |
| Quantity | 6Pack |
| Stopper Number | NS 34/12 |
| Product Line | Bottle |
| Product Type | Weighing Bottle |
| Diameter (Metric) | 30 mm |