missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEF2A Rabbit anti-Human, Mouse, Rat, Clone: S04-2C8, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 295.00 - € 581.00
Specifications
| Antigen | MEF2A |
|---|---|
| Clone | S04-2C8 |
| Dilution | Western Blot, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18063094
|
Novus Biologicals
NBP3-14953-25UL |
25 μg |
€ 295.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18320892
|
Novus Biologicals
NBP3-14953-100UL |
100 μg |
€ 581.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MEF2A Monoclonal antibody specifically detects MEF2A in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| MEF2A | |
| Western Blot, Flow Cytometry, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| 4205 | |
| IgG | |
| Affinity-purified |
| S04-2C8 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| ADCAD1, MADS box transcription enhancer factor 2, polypeptide A (myocyte enhancerfactor 2A), MEF2, myocyte enhancer factor 2A, myocyte-specific enhancer factor 2A, RSRFC4, RSRFC9, Serum response factor-like protein 1 | |
| A synthetic peptide of human MEF2A (Uniprot#: Q02078) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title