missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEF2BNB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561076
This item is not returnable.
View return policy
Description
Immunogen sequence: MEEPEMQLKG KKVTDKFTES VYVLANEPSV ALYRLQEHVR RSLPELAQHK ADMQRWEEQS Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Specifications
| MEF2BNB | |
| Polyclonal | |
| Unconjugated | |
| BORCS8 | |
| 2310045N01Rik; BLOC-1 related complex subunit 8; BLOC-1-related complex subunit 8; Borcs8; MEF2B neighbor; MEF2B neighbor gene protein; MEF2B neighbor gene protein homolog; MEF2BNB; protein MEF2BNB; RIKEN cDNA 2310045N01 gene | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 729991 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96FH0 | |
| BORCS8 | |
| Recombinant protein corresponding to Human MEF2BNB. Recombinant protein control fragment (Product #RP-100562). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction