missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEF2C Antibody (OTI1H5), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72627MFV610
This item is not returnable.
View return policy
Description
MEF2C Monoclonal antibody specifically detects MEF2C in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| MEF2C | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI1H5 | |
| Flow Cytometry | |
| DEL5q14.3, myocyte enhancer factor 2C, myocyte-specific enhancer factor 2C, polypeptide C | |
| Full length human recombinant protein of human MEF2C (NP_002388) produced in HEK293T cell. | |
| 0.1 mL | |
| Neuroscience | |
| 4208 | |
| Store at 4C in the dark. | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion