missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEF2C Monoclonal Antibody (OTI1D1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502501
This item is not returnable.
View return policy
Description
MEF2C is a transcription activator which binds specifically to the MEF2 element present in the regulatory regions of many muscle-specific genes. Controls cardiac morphogenesis and myogenesis, and is also involved in vascular development. Plays an essential role in hippocampal-dependent learning and memory by suppressing the number of excitatory synapses and thus regulating basal and evoked synaptic transmission. Crucial for normal neuronal development, distribution, and electrical activity in the neocortex. Necessary for proper development of megakaryocytes and platelets and for bone marrow B lymphopoiesis. Required for B-cell survival and proliferation in response to BCR stimulation, efficient IgG1 antibody responses to T-cell-dependent antigens and for normal induction of germinal center B cells. May also be involved in neurogenesis and in the development of cortical architecture. Isoform 3 and isoform 4, which lack the repressor domain, are more active than isoform 1 and isoform 2.Specifications
| MEF2C | |
| Monoclonal | |
| Unconjugated | |
| MEF2C | |
| C5DELq14.3, DEL5q14.3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4208 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1D1 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q06413 | |
| MEF2C | |
| Full length human recombit protein of human MEF2C produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction