missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEF2D Antibody (OTI1E11), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72631C
This item is not returnable.
View return policy
Description
MEF2D Monoclonal specifically detects MEF2D in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MEF2D | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MEF2D | |
| Recombinant protein fragment corresponding to amino acids 1-282 of human MEF2D (NP_005911) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1E11 | |
| Western Blot, Immunohistochemistry | |
| DKFZp686I1536, MADS box transcription enhancer factor 2, polypeptide D (myocyte enhancerfactor 2D), MEF2D/DAZAP1 fusion, myocyte enhancer factor 2D, myocyte enhancer factor 2D/deleted in azoospermia associated protein 1 fusionprotein, myocyte-specific enhancer factor 2D | |
| Mouse | |
| Affinity Purified | |
| Phospho Specific | |
| 4209.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction