missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEF2D Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14353-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene is a member of the myocyte-specific enhancer factor 2 (MEF2) family of transcription factors. Members of this family are involved in control of muscle and neuronal cell differentiation and development, and are regulated by class II histone deacetylases. Fusions of the encoded protein with Deleted in Azoospermia-Associated Protein 1 (DAZAP1) due to a translocation have been found in an acute lymphoblastic leukemia cell line, suggesting a role in leukemogenesis. The encoded protein may also be involved in Parkinson disease and myotonic dystrophy. Alternative splicing results in multiple transcript variants.Specifications
| MEF2D | |
| Polyclonal | |
| Unconjugated | |
| MEF2D | |
| MEF2D, myocyte enhancer factor 2D | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17261, 4209, 81518 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O89038, Q14814, Q63943 | |
| Mef2d | |
| MEF2D Fusion Protein Ag5700 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction