missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEGF11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540109
This item is not returnable.
View return policy
Description
This antibody may recoginize two isoforms: isoform 1 (110 kDa) and isoform 2 (101 kDa). Recommended positive controls: mouse eye tissue. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May regulate the mosaic spacing of specific neuron subtypes in the retina through homotypic retinal neuron repulsion. Mosaics provide a mechanism to distribute each cell type evenly across the retina, ensuring that all parts of the visual field have access to a full set of processing elements. [UniProt].
Specifications
| MEGF11 | |
| Polyclonal | |
| Unconjugated | |
| MEGF11 | |
| 2410080H04Rik; D130061K05; KIAA1781; Megf11; multiple EGF like domains 11; multiple EGF-like domains protein 11; multiple EGF-like-domains 11; multiple epidermal growth factor-like domains protein 11; UNQ1949/PRO4432 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 214058, 84465 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.87 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% Thimerosal; pH 7 | |
| A6BM72, Q80T91 | |
| MEGF11 | |
| Recombinant protein encompassing a sequence within the Extracellular domain of human MEGF11. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction