missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEGF8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00 - € 624.00
Specifications
| Antigen | MEGF8 |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18424732
|
Novus Biologicals
NBP2-14228-25ul |
25ul |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18047499
|
Novus Biologicals
NBP2-14228 |
0.1 mL |
€ 624.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MEGF8 Polyclonal specifically detects MEGF8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MEGF8 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 1954 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: PCSPMPRSPEECRRLRTCSECLARHPRTLQPGDGEASTPRCKWCTNCPEGACIGRNGSCTSENDCRINQREVFWAGNCSEAACGAADCEQCTREGKCMWTRQFKRTGETRRILSVQPTYDWTCFSHSLLNVS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| C19orf49, chromosome 19 open reading frame 49, EGFL4MGC120684, EGF-like domain-containing protein 4, EGF-like protein 4, EGF-like-domain, multiple 4, Epidermal growth factor-like protein 4, FLJ22365, HBV pre-s2 binding protein 1, KIAA0817, MGC138147, multiple EGF-like domain protein 4, Multiple EGF-like domains protein 8, multiple EGF-like-domains 8, multiple epidermal growth factor-like domains 8, multiple epidermal growth factor-like domains protein 8, SBP1 | |
| MEGF8 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title