missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Meis homeobox 3 Antibody (OTI3E12), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72642MFV500
This item is not returnable.
View return policy
Description
Meis homeobox 3 Monoclonal antibody specifically detects Meis homeobox 3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
| Meis homeobox 3 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| OTI3E12 | |
| Flow Cytometry | |
| DKFZp547H236, Meis homeobox 3, Meis1, myeloid ecotropic viral integration site 1 homolog 3, Meis1, myeloid ecotropic viral integration site 1 homolog 3 (mouse), Meis1-related protein 2, MRG2homeobox protein Meis3 | |
| Human recombinant protein fragment corresponding to amino acids 1-261 of human MEIS3 (NP_064545) produced in E.coli. | |
| 0.1 mL | |
| Neuroscience | |
| 56917 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction