missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEIS1 Monoclonal Antibody (OTI3F12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809621
This item is not returnable.
View return policy
Description
Acts as a transcriptional regulator of PAX6. Acts as a transcriptional activator of PF4 in complex with PBX1 or PBX2. Required for hematopoiesis, megakaryocyte lineage development and vascular patterning. May function as a cofactor for HOXA7 and HOXA9 in the induction of myeloid leukemias.Specifications
| MEIS1 | |
| Monoclonal | |
| Unconjugated | |
| MEIS1 | |
| Meis homeobox 1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4211 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI3F12 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O00470 | |
| MEIS1 | |
| Human recombit protein fragment corresponding to amino acids 176-390 of human MEIS1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction