Learn More
MEIS1 Monoclonal Antibody (OTI3F12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA809621S
Encompass_Preferred
 View more versions of this product
Product Code. 17582034
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
MEIS1 Monoclonal antibody specifically detects MEIS1 in Human samples. It is validated for Western Blot
Specifications
Specifications
MEIS1
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O00470
Mouse
Affinity chromatography
RUO
4211
-20° C, Avoid Freeze/Thaw Cycles
IgG2b
Western Blot
OTI3F12
Unconjugated
MEIS1
C530044H18Rik; cb224; etID37540.13; Evi8; Homeobox protein Meis1; homeobox protein Meis1b; id:ibd5078; leukemogenic homolog protein; Meis homeobox 1; Meis homeobox 1 b; Meis homeobox 1 S homeolog; meis1; Meis1, myeloid ecotropic viral integration site 1 homolog; meis1.1; meis1.S; MEIS1A; meis1b; MGC43380; myeloid ecotropic viral insertion site-1a protein; myeloid ecotropic viral integration 1; myeloid ecotropic viral integration site 1; transcription factor Meis1.1; wu:fb38b03; wu:fc02e11; wu:fd12a02; WUGSC:H_NH0444B04.1; XMeis1; XMeis1-1 protein; XMeis1-2 protein; XMeis1-4 protein
Human recombinant protein fragment corresponding to amino acids 176-390 of human MEIS1 produced in E.coli.
30 μL