Learn More
MEIS1 Mouse anti-Human, Clone: OTI3H2, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF809623
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Acts as a transcriptional regulator of PAX6. Acts as a transcriptional activator of PF4 in complex with PBX1 or PBX2. Required for hematopoiesis, megakaryocyte lineage development and vascular patterning. May function as a cofactor for HOXA7 and HOXA9 in the induction of myeloid leukemias.Specifications
| MEIS1 | |
| Monoclonal | |
| Unconjugated | |
| MEIS1 | |
| Meis homeobox 1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4211 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3H2 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O00470 | |
| MEIS1 | |
| Human recombit protein fragment corresponding to amino acids 176-390 of human MEIS1 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.