missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEIS1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112605
This item is not returnable.
View return policy
Description
MEIS1 Polyclonal Antibody for Western Blot, IHC (P), IP, ELISA
Homeobox genes, of which the most well-characterized category is represented by the HOX genes, play a crucial role in normal development. In addition, several homeoproteins are involved in neoplasia. This gene encodes a homeobox protein belonging to the TALE family of homeodomain-containing proteins.
Specifications
| MEIS1 | |
| Polyclonal | |
| Unconjugated | |
| MEIS1 | |
| C530044H18Rik; cb224; etID37540.13; Evi8; Homeobox protein Meis1; homeobox protein Meis1b; id:ibd5078; leukemogenic homolog protein; Meis homeobox 1; Meis homeobox 1 b; Meis homeobox 1 S homeolog; meis1; Meis1, myeloid ecotropic viral integration site 1 homolog; meis1.1; meis1.S; MEIS1A; meis1b; MGC43380; myeloid ecotropic viral insertion site-1a protein; myeloid ecotropic viral integration 1; myeloid ecotropic viral integration site 1; transcription factor Meis1.1; wu:fb38b03; wu:fc02e11; wu:fd12a02; WUGSC:H_NH0444B04.1; XMeis1; XMeis1-1 protein; XMeis1-2 protein; XMeis1-4 protein | |
| Rabbit | |
| Protein G | |
| RUO | |
| 17268, 4211 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O00470, Q60954 | |
| MEIS1 | |
| Recombinant Human Homeobox protein Meis1 protein (1-90AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction