missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MEIS1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17298343 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17298343 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17298343 Supplier Invitrogen™ Supplier No. PA5115567

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MEIS1.

Homeobox genes, of which the most well-characterized category is represented by the HOX genes, play a crucial role in normal development. In addition, several homeoproteins are involved in neoplasia. This gene encodes a homeobox protein belonging to the TALE family of homeodomain-containing proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEIS1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene MEIS1
Gene Accession No. O00470
Gene Alias C530044H18Rik; cb224; etID37540.13; Evi8; Homeobox protein Meis1; homeobox protein Meis1b; id:ibd5078; leukemogenic homolog protein; Meis homeobox 1; Meis homeobox 1 b; Meis homeobox 1 S homeolog; meis1; Meis1, myeloid ecotropic viral integration site 1 homolog; meis1.1; meis1.S; MEIS1A; meis1b; MGC43380; myeloid ecotropic viral insertion site-1a protein; myeloid ecotropic viral integration 1; myeloid ecotropic viral integration site 1; transcription factor Meis1.1; wu:fb38b03; wu:fc02e11; wu:fd12a02; WUGSC:H_NH0444B04.1; XMeis1; XMeis1-1 protein; XMeis1-2 protein; XMeis1-4 protein
Gene Symbols MEIS1
Host Species Rabbit
Immunogen A synthesized peptide derived from human MEIS1(Accession O00470), corresponding to amino acid residues L185-G235.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4211
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.