missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEIS2 Antibody (PCRP-MEIS2-2B3), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21014CL1
This item is not returnable.
View return policy
Description
MEIS2 Monoclonal antibody specifically detects MEIS2 in Human samples. It is validated for Flow Cytometry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| MEIS2 | |
| Monoclonal | |
| CoraFluor 1 | |
| Meis (mouse) homolog 2, Meis homeobox 2, Meis1, myeloid ecotropic viral integration site 1 homolog 2 (mouse), Meis1-related gene 1, MGC2820, myeloid ecotropic viral integration site 1 homolog 2, TALE homeobox protein Meis2 | |
| Recombinant fragment of human MEIS2 | |
| 0.1 mL | |
| Vision | |
| 4212 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a κ |
| Flow Cytometry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
| PCRP-MEIS2-2B3 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction