missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEIS2 Antibody (PCRP-MEIS2-2B3), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21014G
This item is not returnable.
View return policy
Description
MEIS2 Monoclonal antibody specifically detects MEIS2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| MEIS2 | |
| Monoclonal | |
| DyLight 488 | |
| Meis (mouse) homolog 2, Meis homeobox 2, Meis1, myeloid ecotropic viral integration site 1 homolog 2 (mouse), Meis1-related gene 1, MGC2820, myeloid ecotropic viral integration site 1 homolog 2, TALE homeobox protein Meis2 | |
| Recombinant fragment of human MEIS2 | |
| 0.1 mL | |
| Vision | |
| 4212 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| PCRP-MEIS2-2B3 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction