missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEIS2 Antibody (PCRP-MEIS2-2B3), Novus Biologicals™
Mouse Monoclonal Antibody
€ 275.00 - € 530.00
Specifications
| Antigen | MEIS2 |
|---|---|
| Clone | PCRP-MEIS2-2B3 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, Immunoprecipitation 1-2 μg per 100-500 μg of protein or 1ml of lysate, Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18632215
|
Novus Biologicals
NBP3-20520-20ug |
20 μg |
€ 275.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18629584
|
Novus Biologicals
NBP3-20520-100ug |
100 μg |
€ 530.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MEIS2 Monoclonal antibody specifically detects MEIS2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| MEIS2 | |
| 0.2 mg/ml | |
| Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Vision | |
| 10mM PBS with 0.05% BSA | |
| 4212 | |
| IgG2a κ | |
| Protein A or G purified |
| PCRP-MEIS2-2B3 | |
| Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, Immunoprecipitation 1-2 μg per 100-500 μg of protein or 1ml of lysate, Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Meis (mouse) homolog 2, Meis homeobox 2, Meis1, myeloid ecotropic viral integration site 1 homolog 2 (mouse), Meis1-related gene 1, MGC2820, myeloid ecotropic viral integration site 1 homolog 2, TALE homeobox protein Meis2 | |
| Recombinant fragment of human MEIS2 | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title