missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MEIS2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16599881
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16599881 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16599881 Supplier Invitrogen™ Supplier No. PA597053

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: AYSPEGQPMG SFVLDGQQHM GIRPAGLQSM PGDYVSQGGP MGMSMAQPSY TPPQMTPHPT QLRHGPPMHS YLPSHPHHPA MMMHGGPPTH PGMTMSAQSP TMLNSVDPNV GGQVMDIHAQ; Positive Samples: Mouse kidney, Mouse heart, Rat kidney; Cellular Location: Cytoplasm, Nucleus, perinuclear region.

This gene encodes a homeobox protein belonging to the TALE family of homeodomain-containing proteins. TALE homeobox proteins are highly conserved transcription regulators, and several members have been shown to be essential contributors to developmental programs. Multiple transcript variants encoding distinct isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEIS2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.42 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MEIS2
Gene Accession No. P97367
Gene Alias A430109D20Rik; Cbp/p300-interacting transactivator with Glu/Asp-rich carboxy-terminal domain 2; homeobox protein Meis2; HsT18361; Meis homeobox 2; Meis homolog 2; Meis1, myeloid ecotropic viral integration site 1 homolog 2; Meis1-related gene 1; meis1-related protein 1; MEIS2; Meis2 homeobox; MGC2820; MRG1; myeloid ecotropic viral integration site-related gene 1; OTTHUMP00000160209; Stra10; TALE homeobox protein Meis2
Gene Symbols MEIS2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 351-470 of human MEIS2 (NP_7337761).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17536, 311311
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.