missing translation for 'onlineSavingsMsg'
Learn More

MEIS3 Monoclonal Antibody (OTI3E12), TrueMAB™, OriGene

Product Code. p-200016409
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17567962 30 μL 30µL
17547962 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17567962 Supplier Origene Technologies Supplier No. TA800268S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MEIS3 Monoclonal antibody specifically detects MEIS3 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MEIS3
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3E12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MEIS3
Gene Accession No. Q99687
Gene Alias AI573393; Homeobox protein Meis3; Meis homeobox 3; Meis homeobox 3 pseudogene 1; Meis1, myeloid ecotropic viral integration site 1 homolog 3; meis1-related protein 2; Meis3; MEIS3P1; Mrg2; myeloid ecotropic viral integration site-related gene 2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-261 of human MEIS3 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56917
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.