missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEK1, p Thr292 Antibody (SR1773), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | MEK1 |
|---|---|
| Clone | SR1773 |
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18624035
|
Novus Biologicals
NBP3-21559-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633965
|
Novus Biologicals
NBP3-21559-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MEK1 Monoclonal antibody specifically detects MEK1 in Human samples. It is validated for Western BlotSpecifications
| MEK1 | |
| Western Blot 1:500-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| dual specificity mitogen-activated protein kinase kinase 1, ERK activator kinase 1, MAP kinase kinase 1, MAPK/ERK kinase 1, MAPKK1, MEK1MAPKK 1, mitogen-activated protein kinase kinase 1, MKK1MEK 1, PRKMK1EC 2.7.12.2, protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1) | |
| A synthesized peptide derived from human phospho-MEK1 [Thr292] (Uniprot #: Q02750 ) | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
| SR1773 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Autophagy, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Phosphatase, Signal Transduction, Tyrosine Kinases | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 5604 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title