missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEK1 Recombinant Rabbit Monoclonal Antibody (BLR096G)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544384
This item is not returnable.
View return policy
Description
MEK1 Recombinant Monoclonal Antibody for Western Blot, Flow, IP
MAP2K1 (MEK1) kinase is a dual-specificity protein kinase that functions in mitogen-activated protein kinase (MAPK) cascades, controlling cell growth and differentiation. MAP2K1 is activated by a wide variety of growth factors and cytokines and also by membrane-dependent depolarization and calcium influx. Mek1 is activated by phosphorylation of serine 218 and 222 residues by Raf1. It is known to be involved in the signaling during stress activate response, apoptosis and proliferative induction by cytokines. Mutations in the gene can lead to cardiofaciocutaneous syndrome.
Specifications
| MEK1 | |
| Recombinant Monoclonal | |
| 100 μg/mL | |
| BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6 | |
| Q02750 | |
| Map2k1 | |
| Peptide representing a region between residue 343 and the C-terminus (residue 393) of human dual specificity mitogen-activated protein kinase kinase 1 (MEK1). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| BLR096G | |
| Unconjugated | |
| Map2k1 | |
| CFC3; dual specificity mitogen activated protein kinase kinase 1; dual specificity mitogen-activated protein kinase kinase 1; EC 2.7.12.2; ERK activator kinase 1; kinase MEK1; MAP kinase kinase 1; MAP kinase kinase or Erk Kinase, Dual specificity mitogen-activated protein kinase kinase, involved in ras mediated vulval induction, LEThal LET-537 (42.8 kD) (mek-2); MAP kinase/Erk kinase 1; Map2k1; map2k1.L; MAPK/ERK kinase 1; MAPKK 1; MAPKK1; MEK 1; Mek1; MEK2; mek-2; MEKK1; mitogen activated protein kinase kinase 1; mitogen-activated protein kinase kinase 1; mitogen-activated protein kinase kinase 1 L homeolog; MKK (Thr292); MKK (Thr386); MKK1; MKK2; MP2K1; Prkmk1; PRKMK2; protein kinase, mitogen activated, kinase 1, p45; protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1); XELAEV_18018447mg | |
| Rabbit | |
| Purified | |
| RUO | |
| 5604 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction