missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587392
This item is not returnable.
View return policy
Description
Positive Samples: Mouse brain; Cellular Location: Cytoplasm, Membrane, Peripheral membrane protein Immunogen sequence: MLARRKPVLP ALTINPTIAE GPSPTSEGAS EANLVDLQKK LEELELDEQQ KKRLEAFLTQ KAKVGELKDD DFERISELGA GNGGVVTKVQ HRPSGLIMAR.
MAP2K2 (MEK2) is a dual specificity serine/threonine kinase that is known to play a critical role in mitogen growth factor signal transduction. Mitogen activated protein kinase 2 (MEK2 or MAPKK2) is a member of a family of tyrosine/threonine protein kinases that activate the ERK1 and 2 and MAPK enzymes by phosphorylating both residues within the threonine/glutamate/tyrosine (TEY) motif in the activation loop. MEK1 and 2 are also activated by dual phosphorylation, which occurs on serine 218 and 222, in the activation loop of the MEK. Threonine 292 of MEK1 is phosphorylated by ERK 2, which serves as a negative feedback loop by suppressing activation of MEK1.
Specifications
| MEK2 | |
| Polyclonal | |
| Unconjugated | |
| MAP2K2 | |
| AA589381; CFC4; Dual specificity mitogen-activated protein kinase kinase 2; ERK activator kinase 2; ERK kinase 2; FLJ26075; MAP kinase kinase 2; MAP kinase/Erk kinase; MAP2K1; MAP2K2; MAPK; MAPK/ERK kinase 1; MAPK/ERK kinase 2; MAPKK 1; MAPKK 2; MAPKK2; MEK 1; MEK 2; MEK-1; Mek2; MEK-2; mitogen activated protein kinase kinase 2; mitogen-activated protein kinase kinase 2; mitogen-activated protein kinase kinase 2, p45; MK2; MKK1; Mkk2; OTTHUMP00000165827; PRKMK1; PRKMK2; protein kinase, mitogen activated, kinase 2, p45 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 26396 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.81 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q63932 | |
| MAP2K2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MEK2 (NP_1095871). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction