missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEK2 Recombinant Rabbit Monoclonal Antibody (BLR079G)
Description
Epitope retrieval with citrate buffer pH6.0 is recommended for FFPE cell sections.
MAP2K2 (MEK2) is a dual specificity serine/threonine kinase that is known to play a critical role in mitogen growth factor signal transduction. Mitogen activated protein kinase 2 (MEK2 or MAPKK2) is a member of a family of tyrosine/threonine protein kinases that activate the ERK1 and 2 and MAPK enzymes by phosphorylating both residues within the threonine/glutamate/tyrosine (TEY) motif in the activation loop. MEK1 and 2 are also activated by dual phosphorylation, which occurs on serine 218 and 222, in the activation loop of the MEK. Threonine 292 of MEK1 is phosphorylated by ERK 2, which serves as a negative feedback loop by suppressing activation of MEK1.
Specifications
Specifications
| Antigen | MEK2 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | BLR079G |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6 |
| Gene | MAP2K2 |
| Gene Accession No. | P36507, Q63932 |
| Gene Alias | AA589381; CFC4; Dual specificity mitogen-activated protein kinase kinase 2; ERK activator kinase 2; ERK kinase 2; FLJ26075; MAP kinase kinase 2; MAP kinase/Erk kinase; MAP2K1; MAP2K2; MAPK; MAPK/ERK kinase 1; MAPK/ERK kinase 2; MAPKK 1; MAPKK 2; MAPKK2; MEK 1; MEK 2; MEK-1; Mek2; MEK-2; mitogen activated protein kinase kinase 2; mitogen-activated protein kinase kinase 2; mitogen-activated protein kinase kinase 2, p45; MK2; MKK1; Mkk2; OTTHUMP00000165827; PRKMK1; PRKMK2; protein kinase, mitogen activated, kinase 2, p45 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?