missing translation for 'onlineSavingsMsg'
Learn More

MEK5 Monoclonal Antibody (2C2)
GREENER_CHOICE

Product Code. 15548832
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15548832 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15548832 Supplier Invitrogen Supplier No. MA519712

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MEK5 Monoclonal Antibody (2C2)

MEK5 Monoclonal Antibody for Western Blot, ICC/IF, ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen MEK5
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2C2
Concentration 0.2 to 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene MAP2K5
Gene Accession No. Q13163
Gene Alias AI324775; AI428457; Dual specificity mitogen-activated protein kinase kinase 5; HsT17454; MAP kinase kinase 5; MAP kinase kinase MEK5b; MAP2K5; MAPK/ERK kinase 5; MAPK/ERK kinese 5; MAPKK 5; Mapkk5; MEK 5; MEK5; mitogen activated protein kinase kinase 5; mitogen-activated protein kinase kinase 5; Mkk5; PRKMK5
Gene Symbols MAP2K5
Host Species Mouse
Immunogen MAP2K5 (AAH08838, 1 a.a. ~ 120 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5607
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.