missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MEK5 Recombinant Rabbit Monoclonal Antibody (7B7O10)
GREENER_CHOICE

Product Code. 17175833
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17175833 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17175833 Supplier Invitrogen™ Supplier No. MA535172

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: DVYRKMPEHV LGRIAVAVVK GLTYLWSLKI LHRDVKPSNM LVNTRGQVKL CDFGVSTQLV NSIAKTYVGT NAYMAPERIS GEQYGIHSDV WSLGISFMEL A.

MAP2K5 (MEK5) serine/threonine kinase (mitogen-activated protein kinase 5) or MEK5 is a dual specificity protein kinase that belongs to the MAP kinase family which specifically interacts with and activates MAPK7/ERK5. MAP2K5 itself can be phosphorylated and activated by MAP3K3/MEKK3, as well as by atypical protein kinase C isoforms (aPKCs). The signal cascade mediated by MAP2K5 is involved in growth factor stimulated cell proliferation and muscle cell differentiation. The rs3743354 polymorphism in the MAP2K5 promoter may affect the risk of developing colorectal cancer (CRC).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MEK5
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 7B7O10
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene MAP2K5
Gene Accession No. Q13163, Q62862, Q9WVS7
Gene Alias AI324775; AI428457; Dual specificity mitogen-activated protein kinase kinase 5; HsT17454; MAP kinase kinase 5; MAP kinase kinase MEK5b; MAP2K5; MAPK/ERK kinase 5; MAPK/ERK kinese 5; MAPKK 5; Mapkk5; MEK 5; MEK5; mitogen activated protein kinase kinase 5; mitogen-activated protein kinase kinase 5; Mkk5; PRKMK5
Gene Symbols MAP2K5
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 250-350 of human MEK5 (Q13163).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23938, 29568, 5607
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.