missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MEKK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA554443
This item is not returnable.
View return policy
Description
Immunogen sequence: IAFGQSMDLH YTNNELVIPL TTQDDLDKAV ELLDRSIHMK SLKILLVING STQATNLEPL PSLEDLDNTV FGAERKKRLS IIGPTSRDRS SPPPGYIPDE LHQVARNGSF TSINSEGEFI PESMDQM Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.
MAP3K2 (MEKK2) is a serine/threonine kinase that activates kinases involved in the MAP kinase signaling pathway. This kinase preferentially activates other kinases involved in the MAP kinase signaling pathway. This kinase has been shown to directly phosphorylate and activate Ikappa B kinases, and thus plays a role in NF-kappa B signaling pathway. This kinase has also been found to bind and activate protein kinase C-related kinase 2, which suggests its involvement in a regulated signaling process.
Specifications
| MEKK2 | |
| Polyclonal | |
| Unconjugated | |
| Map3k2 | |
| 2.7.11.25; 9630061B06Rik; AI585793; MAP/ERK kinase kinase 2; MAP3K2; MAPK/ERK kinase kinase 2; MAPKKK2; MEK kinase 2; MEKK 2; MEKK2; MEKK2B; mitogen activated protein kinase kinase kinase 2; mitogen-activated protein kinase kinase kinase 2; protein kinase MEKK2b | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10746 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y2U5 | |
| Map3k2 | |
| Recombinant protein corresponding to Human MEKK2. Recombinant protein control fragment (Product #RP-102059). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction