missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melan-A/MART-1 Antibody (DT101 + BC199), Novus Biologicals™
Mouse Monoclonal Antibody
€ 276.00 - € 550.00
Specifications
| Antigen | Melan-A/MART-1 |
|---|---|
| Clone | DT101 + BC199 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18440722
|
Novus Biologicals
NBP2-34246-0.02mg |
0.02 mg |
€ 276.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18421622
|
Novus Biologicals
NBP2-34246-0.1mg |
0.1 mg |
€ 550.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Melan-A/MART-1 Monoclonal antibody specifically detects Melan-A/MART-1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Melan-A/MART-1 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Biology, Cellular Markers, Immunology | |
| 10 mM PBS with 0.05% BSA | |
| Antigen LB39-AA, Antigen SK29-AA, Mart 1 Melan A, MART1MART-1, melan-A, melanoma antigen recognized by T-cells 1, Protein Melan-A | |
| Recombinant hMelan-A/MART-1 protein (DT101 & BC199) (Uniprot: Q16655) | |
| Primary | |
| Store at 4C. |
| DT101 + BC199 | |
| Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q16655, Q16655, Q16655 | |
| 2315 | |
| IgG2b λ, IgG2b κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title